missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenyl isothiocyanate, 95%, ACROS Organics™

Brand:  Acros Organics 441780050

Additional Details : CAS : 33904-03-9 Weight : 0.05500kg Transport : UN number : 3261 Chem class : 8 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 11965771

  • € 51.23 / 5g


In stock, ready to ship: 636
Add to basket

Description

Description

Specifications

Chemical Identifiers

33904-03-9
195.24
2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
1-isothiocyanato-2,4-dimethoxybenzene
C9H9NO2S
CNXSCEPTSXPBTP-UHFFFAOYSA-N
2736202
COC1=CC(=C(C=C1)N=C=S)OC

Specifications

2, 4-Dimethoxyphenyl isothiocyanate
Authentic
Yellow
5g
94.0
94% min. (GC)
(CH3O)2C6H3NCS
CNXSCEPTSXPBTP-UHFFFAOYSA-N
1-isothiocyanato-2,4-dimethoxybenzene
2736202
Powder
113°C
166.0°C to 167.0°C (6.0mmHg)
54.0°C to 55.0°C
33904-03-9
100.0
C9H9NO2S
2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
COC1=CC(=C(C=C1)N=C=S)OC
195.24
195.24
95%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,4-Dimethoxyphenyl isothiocyanate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates