missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Isopropylphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64123.03

Additional Details : CAS : 67273-56-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15441179

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

67273-56-7
236.318
KZLWMCPIYIDTTK-UHFFFAOYSA-N
735512
CC(C)C1=CC=C(C=C1)C2=NC3=CC=CC=C3N2
C16H16N2
MFCD05042196
2-4-isopropylphenyl-1h-benzimidazole, 2-4-methylethyl phenyl benzimidazole, 2-4-propan-2-ylphenyl-1h-benzimidazole, 1h-benzimidazole, 2-4-1-methylethyl phenyl, 2-4-isopropylphenyl-1h-1,3-benzodiazole
2-(4-propan-2-ylphenyl)-1H-benzimidazole

Specifications

2-(4-Isopropylphenyl)benzimidazole
67273-56-7
MFCD05042196
KZLWMCPIYIDTTK-UHFFFAOYSA-N
2-(4-propan-2-ylphenyl)-1H-benzimidazole
735512
Solid
1g
C16H16N2
2-4-isopropylphenyl-1h-benzimidazole, 2-4-methylethyl phenyl benzimidazole, 2-4-propan-2-ylphenyl-1h-benzimidazole, 1h-benzimidazole, 2-4-1-methylethyl phenyl, 2-4-isopropylphenyl-1h-1,3-benzodiazole
CC(C)C1=CC=C(C=C1)C2=NC3=CC=CC=C3N2
236.318
236.32
95%
Videos
Documents