missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-2-phenylpiperazin-1-yl)pyridine-3-carbonitrile, Mikromol

Product Code. 30460552
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30460552 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460552 Supplier Mikromol Supplier No. MM1005.140025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(4-Methyl-2-phenylpiperazin-1-yl)pyridine-3-carbonitrile, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(3-Cyano-2-pyridyl)-4-methyl-2-phenylpiperazine
CAS 61337-88-0
Analyte or Component Names 1-(3-Cyano-2-pyridyl)-4-methyl-2-phenylpiperazine
Shipping Condition Room Temperature
Molecular Formula C17 H18 N4
Certifications/Compliance ISO 9001
Synonym 3-Pyridinecarbonitrile, 2-(4-methyl-2-phenyl-1-piperazinyl)-, 2-(4-Methyl-2-phenyl-1-piperazinyl)-3-pyridinecarbonitrile, 1-(3-Cyano-2-pyridyl)-4-methyl-2-phenylpiperazine, 2-(4-Methyl-2-phenylpiperazin-1-yl)-3-cyanopyridine, 2-(4-Methyl-2-phenylpiperazin-1-yl)pyridine-3-carbonitrile
InChI Formula InChI=1S/C17H18N4/c1-20-10-11-21(17-15(12-18)8-5-9-19-17)16(13-20)14-6-3-2-4-7-14/h2-9,16H,10-11,13H2,1H3
Solution Type Neat
SMILES CN1CCN(C(C1)c2ccccc2)c3ncccc3C#N
IUPAC Name 2-(4-methyl-2-phenylpiperazin-1-yl)pyridine-3-carbonitrile
Molecular Weight (g/mol) 278.35
Quantity 25 mg
Formula Weight 278.1531 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.