missing translation for 'onlineSavingsMsg'
Learn More

{2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine, 97%, Thermo Scientific™

Catalog Number p-3175648
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
879896-50-1
C13H21N3
219.332
MFCD06797802
OKGWICVOLZFMPE-UHFFFAOYSA-N
2-4-methylpiperazin-1-ylmethyl benzylamine, 1-2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazinyl methyl phenyl methylamine, 2-4-methyl-1-piperazinyl methyl-benzenemethanamine, 2-4-methylpiperazin-1-yl methyl phenyl methylamine, benzenemethanamine,2-4-methyl-1-piperazinyl methyl
16228064
[2-[(4-methylpiperazin-1-yl)methyl]phenyl]methanamine
CN1CCN(CC1)CC2=CC=CC=C2CN
This item is not returnable. View return policy
879896-50-1
C13H21N3
219.332
MFCD06797802
OKGWICVOLZFMPE-UHFFFAOYSA-N
2-4-methylpiperazin-1-ylmethyl benzylamine, 1-2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazinyl methyl phenyl methylamine, 2-4-methyl-1-piperazinyl methyl-benzenemethanamine, 2-4-methylpiperazin-1-yl methyl phenyl methylamine, benzenemethanamine,2-4-methyl-1-piperazinyl methyl
16228064
[2-[(4-methylpiperazin-1-yl)methyl]phenyl]methanamine
CN1CCN(CC1)CC2=CC=CC=C2CN
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 219.33
Percent Purity 97%
Chemical Name or Material {2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'