Learn More
2-(4-Morpholinyl)-5-nitropyridine, 95%, Thermo Scientific Chemicals
Product Code. 15447397
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447397 5 g 5g
15437397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15447397 Supplier Thermo Scientific Alfa Aesar Supplier No. H34452.06
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26820-62-2
Molecular Formula C9H11N3O3
Molecular Weight (g/mol) 209.21
MDL Number MFCD00023308
InChI Key FYJXIPHXFGFEGT-UHFFFAOYSA-N
Synonym 4-5-nitropyridin-2-yl morpholine, 4-5-nitro-2-pyridinyl morpholine, 4-5-nitro-2-pyridyl morpholine, 4-5-nitro-pyridin-2-yl-morpholine, morpholine, 4-5-nitro-2-pyridyl, 2-morpholino-5-nitropyridine, morpholine, 4-5-nitro-2-pyridinyl, pubchem17157, acmc-1cizi, cbmicro_021094
PubChem CID 96872
IUPAC Name 4-(5-nitropyridin-2-yl)morpholine
SMILES [O-][N+](=O)C1=CC=C(N=C1)N1CCOCC1
Specifications
Melting Point 142°C to 144°C
Quantity 5 g
Beilstein 0225480
Formula Weight 209.2
Percent Purity 95%
Chemical Name or Material 2-(4-Morpholinyl)-5-nitropyridine