Learn More
2,4-Pteridinediamine-6-methanol Hydrobromide, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Pteridinediamine-6-methanol','Hydrobromide |
| CAS | 57963-59-4 |
| Molecular Formula | C7 H8 N6 O . Br H |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C7H8N6O.BrH/c8-5-4-6(13-7(9)12-5)10-1-3(2-14)11-4;/h1,14H,2H2,(H4,8,9,10,12,13);1H |
| SMILES | Br.Nc1nc(N)c2nc(CO)cnc2n1 |
| IUPAC Name | (2,4-diaminopteridin-6-yl)methanol;hydrobromide |
| Molecular Weight (g/mol) | 273.09 |
| Formula Weight | 272.0021 |