missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-3881278
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10780734 1 g 1g
10650135 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10780734 Supplier Thermo Scientific Maybridge Supplier No. CD11334DA
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 152457-95-9
Molecular Formula C10H7ClF6O4S
Molecular Weight (g/mol) 372.66
MDL Number MFCD00052841
InChI Key UCHAULDYIPVTOP-UHFFFAOYSA-N
Synonym 2,5-bis 2,2,2-trifluoroethoxy benzenesulfonyl chloride, 2,5-bis 2,2,2-trifluoroethoxy benzene-1-sulfonyl chloride, 2,5-bis 2,2,2-trifluoroethoxy benzenesulfonylchloride, 2,5-bis 2,2,2-trifluoroethoxy benzenesulphonyl chloride, 2,5-bis 2,2,2-trifluoroethoxy benzenesulfonyl, 2,5-bis-2,2,2-trifluoroethoxy benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-bis 2,2,2-trifluoroethoxy, 2,5-bis 2,2,2-trifluoroethoxy benzenesulphonyl chloride, tech., pubchem5052, acmc-1c3ia
PubChem CID 2736077
SMILES FC(F)(F)COC1=CC(=C(OCC(F)(F)F)C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2,5-bis(2,2,2-trifluoroethoxy)benzenesulfonyl chloride
Formula Weight 372.67
Percent Purity 97%
Chemical Name or Material 2,5-Di(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.