missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-3-nitropyridine, ≥97%, Thermo Scientific™

Catalog Number p-7055643
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
21427-62-3
C5H2Cl2N2O2
192.983
MFCD06658963
OBUGJYJQJWMOQO-UHFFFAOYSA-N
8063331
2,5-dichloro-3-nitropyridine
C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
21427-62-3
C5H2Cl2N2O2
192.983
MFCD06658963
OBUGJYJQJWMOQO-UHFFFAOYSA-N
8063331
2,5-dichloro-3-nitropyridine
C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 41°C to 45°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN2811
Beilstein 147539
Synonym 2,5-dichloro-3-nitro-pyridine, pyridine, 2,5-dichloro-3-nitro, 2.5-dichloro-3-nitropyridine, 2,5-dichloro-3-nitro pyridine, pubchem2267, acmc-1cdlk, 2,5dichloro-3-nitropyridine, ksc495e7t, 2,5 dichloro-3-nitropyridine, 2,5 dichloro-3-nitro pyridine
Formula Weight 192.98
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 2,5-Dichloro-3-nitropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'