Learn More
2,5-Dimethoxytetrahydro-3-furancarboxaldehyde, Technical Grade, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Dimethoxytetrahydro-3-furancarboxaldehyde |
| CAS | 50634-05-4 |
| Molecular Formula | C7H12O4 |
| Quantity | 5 mg |
| Synonym | 2,5-Dimethoxy-3-formyltetrahydrofuran, 2,5-Dimethoxy-tetrahydro-furan-4-carboxaldehyde, 3-Formyl-2,5-dimethoxytetrahydrofuran, Tetrahydro-2,5-dimethoxy-3-furancarboxaldehyde |
| InChI Formula | InChI=1S/C7H12O4/c1-9-6-3-5(4-8)7(10-2)11-6/h4-7H,3H2,1-2H3 |
| SMILES | O=CC1CC(OC)OC1OC |
| IUPAC Name | 2,5-dimethoxytetrahydrofuran-3-carbaldehyde |
| Molecular Weight (g/mol) | 160.17 |
| Formula Weight | 160.07 |