missing translation for 'onlineSavingsMsg'
Learn More

(2,5-Dimethyl-3-furyl)methylamine, 97%, Thermo Scientific™

Catalog Number 10712103
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
306934-85-0
C7H11NO
125.171
MFCD02677701
AGQXLVABIKZJJG-UHFFFAOYSA-N
2,5-dimethyl-3-furyl methylamine, 2,5-dimethylfuran-3-yl methanamine, 3-furanmethanamine,2,5-dimethyl, 3-aminomethyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl methanamine, 2,5-dimethyl-3-furanmethanamine, 2,5-dimethylfur-3-yl methylamine, 2,5-dimethylfuran-3-yl methyl amine, sz9
2776162
(2,5-dimethylfuran-3-yl)methanamine
CC1=CC(=C(O1)C)CN
This item is not returnable. View return policy
306934-85-0
C7H11NO
125.171
MFCD02677701
AGQXLVABIKZJJG-UHFFFAOYSA-N
2,5-dimethyl-3-furyl methylamine, 2,5-dimethylfuran-3-yl methanamine, 3-furanmethanamine,2,5-dimethyl, 3-aminomethyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl methanamine, 2,5-dimethyl-3-furanmethanamine, 2,5-dimethylfur-3-yl methylamine, 2,5-dimethylfuran-3-yl methyl amine, sz9
2776162
(2,5-dimethylfuran-3-yl)methanamine
CC1=CC(=C(O1)C)CN
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 125.17
Percent Purity 90%
Chemical Name or Material (2, 5-Dimethyl-3-furyl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'