Learn More
2,5-Dimethylhexane, 99%
Product Code. 11489140
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11489140
Brand: Thermo Scientific Alfa Aesar A15221.14
Request Bulk or Custom Format
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
CAS: 592-13-2 | C8H18 | 114.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 592-13-2
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00008951
InChI Key UWNADWZGEHDQAB-UHFFFAOYSA-N
Synonym biisobutyl, hexane, 2,5-dimethyl, diisobutyl, 2,5-dimethyl hexane, hexane,5-dimethyl, acmc-209mbd, 2,5-dimethylhexane, 2,5-dimethylhexane 5g, hexane, 2,5-dimethyl-8ci 9ci
PubChem CID 11592
IUPAC Name 2,5-dimethylhexane
SMILES CC(C)CCC(C)C
Specifications
Density 0.694
Boiling Point 107°C to 108°C
Flash Point 26°C (80°F)
Refractive Index 1.392
Quantity 25 g
UN Number UN3295
Beilstein 1696877
Formula Weight 114.23
Percent Purity 99%
Chemical Name or Material 2,5-Dimethylhexane