missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one, 97%, Thermo Scientific™

Product Code. 11429926
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429926 5 g 5g
11419926 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429926 Supplier Thermo Scientific Alfa Aesar Supplier No. B20888.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98488-10-9
Molecular Formula C8H9N3O
Molecular Weight (g/mol) 163.18
MDL Number MFCD08729601
InChI Key LGQMDCIHBYTQJU-UHFFFAOYSA-N
Synonym 2,5-dimethylpyrazolo 1,5-a pyrimidin-7-ol, 2,5-dimethylpyrazolo 1,5-a pyrimidin-7 4h-one, 2,5-dimethyl-4h-pyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethyl-4,7-dihydropyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethyl-1h-pyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethylpyrazolo 1,5-a pyrimidin-7-one, 2,5-dimethyl-4h,7h-pyrazolo 1,5-a pyrimidin-7-one, acmc-20aoa7, maybridge1_004141
PubChem CID 736199
IUPAC Name 2,5-dimethyl-1H-pyrazolo[1,5-a]pyrimidin-7-one
SMILES CC1=CC2=NC(C)=CC(=O)N2N1

Specifications

Melting Point 252°C to 254°C
Quantity 5 g
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.