missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Diphenyloxazole, Pure, Scintillation Counting, Fisher Chemical™
Click to view available options
Quantity:
100 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Melting Point | 70°C |
Color | White |
Boiling Point | 360°C |
Packaging | Plastic powder jar |
Quantity | 100 g |
Formula Weight | 221.26g/mol |
Physical Form | Powder |
Safety and Handling
Product Identifier
- 2,5-Diphenyloxazole
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'