missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxytetracycline Glucose Yeast Extract Agar Base (Dehydrated)

Product Code. 12791550
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.50kg
500g
This item is not returnable. View return policy

Product Code. 12791550

Brand: Thermo Scientific™ CM0545R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selectively enumerate yeasts and molds from foodstuffs using this selective medium.

Selectively enumerate yeasts and molds from foodstuffs with Thermo Scientific™ Oxoid™ Oxytetracycline Glucose Yeast Extract Agar Base (Dehydrated). 1,2. The medium is based upon the formulation described by Mossel et al.3 which utilizes oxytetracycline selectivity to provide increased counts of yeasts and molds compared to other media that utilize a low pH as a selective agent. Improve the recovery of physically stressed yeast cells on this neutral pH medium4.

Use Oxytetracycline Glucose Yeast Extract Agar for selection and enumeration of yeasts and molds from foodstuffs.

  • Increased colony count of yeasts and molds compared to that of low pH media.
  • Improved recovery of physically stressed yeast cells.

The choice of suitable medium for enumeration of yeast and moulds is greatly dependent on the nature of the foodstuffs under investigation and the organisms that occur on them5. Oxytetracycline Glucose Yeast Extract Agar Base remains bacteriostatic when inoculated with not greater than 1 mL of a 10-1 dilution of foods and subsequently incubated for not greater than 5 days at 25°C as is the customary practice in food mycology2.

For isolation of psychrotrophic yeasts from chilled proteinaceous foods a combination of oxytetracycline and gentamicin is effective6.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, ISO, NMKL.
Form Powder
Product Type Agar
Quantity 2.5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.