missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wilkinson’s Catalyst Sulfate Sodium Salt, TRC
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
Specifications
Specifications
| Chemical Name or Material | Wilkinson’s Catalyst Sulfate Sodium Salt |
| Analyte or Component Names | Wilkinson’s Catalyst Sulfate Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C54 H36 Cl Na9 O27 P3 Rh S9 |
| Synonym | Chlorotris(triphenylphosphine)rhodium Sulfate Sodium Salt, Rhodium tris(triphenylphosphine) Chloride Sulfate Sodium Salt, Tris(triphenylphosphine)chlororhodium Sulfate Sodium Salt, Tris(triphenylphosphine)rhodium(I) Chloride Sulfate Sodium Salt |
| InChI Formula | InChI=1S/3C18H15O9PS3.ClH.9Na.Rh/c3*19-29(20,21)16-7-1-13(2-8-16)28(14-3-9-17(10-4-14)30(22,23)24)15-5-11-18(12-6-15)31(25,26)27;;;;;;;;;;;/h3*1-12H,(H,19,20,21)(H,22,23,24)(H,25,26,27);1H;;;;;;;;;;/q;;;;10*+1/p-10 |
| Solution Type | Neat |
| SMILES | [Na]OS(=O)(=O)c1ccc(cc1)[P](c2ccc(cc2)S(=O)(=O)O[Na])(c3ccc(cc3)S(=O)(=O)O[Na])[Rh](Cl)([P](c4ccc(cc4)S(=O)(=O)O[Na])(c5ccc(cc5)S(=O)(=O)O[Na])c6ccc(cc6)S(=O)(=O)O[Na])[P](c7ccc(cc7)S(=O)(=O)O[Na])(c8ccc(cc8)S(=O)(=O)O[Na])c9ccc(cc9)S(=O)(=O)O[Na] |
| IUPAC Name | chloro-tris[tris(4-sodiooxysulfonylphenyl)-λ4-phosphanyl]rhodium |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction