missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caffeine-d3 (1-methyl-d3), TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Caffeine-d3 |
| CAS | 26351-03-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8H7D3N4O2 |
| Quantity | 2.5 mg |
| Synonym | 1H-Purine-2,6-dione, 3,7-dihydro-3,7-dimethyl-1-(methyl-d3)-, Caffeine-α1,α1,α1-d3 (8CI), 3,7-Dihydro-3,7-dimethyl-1-(methyl-d3)-1H-purine-2,6-dione, 1-Trideuteromethyl-3,7-dimethylxanthine, Caffeine-d3, 3,7-Dihydro-3,7-dimethyl-1-(methyl-d3)-1H-purine-2,6-dione, Caffeine-D3 (1-methyl-D3) |
| InChI Formula | InChI=1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i3D3 |
| SMILES | O=C(N(C)C1=C2N(C)C=N1)N(C([2H])([2H])[2H])C2=O |
| IUPAC Name | 3,7-dihydro-3,7-dimethyl-1-(methyl-d3)-1H-purine-2,6-dione |
| Molecular Weight (g/mol) | 197.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction