Learn More
2,6-Diaminopurine Hemisulfate, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Diaminopurine, Hemisulfate Salt |
| CAS | 69369-16-0 |
| Molecular Formula | 2 C5 H6 N6 . H2 O4 S |
| Quantity | 10 g |
| Synonym | 1H-Purine-2,6-diamine, sulfate (2:1) (9CI), 2,6-Diaminopurine hemisulfate |
| InChI Formula | InChI=1S/2C5H6N6.H2O4S/c2*6-3-2-4(9-1-8-2)11-5(7)10-3;1-5(2,3)4/h2*1H,(H5,6,7,8,9,10,11);(H2,1,2,3,4) |
| SMILES | Nc1nc(N)c2[nH]cnc2n1.Nc3nc(N)c4[nH]cnc4n3.OS(=O)(=O)O |
| IUPAC Name | 7H-purine-2,6-diamine;sulfuric acid |
| Molecular Weight (g/mol) | 398.36 |
| Formula Weight | 398.0982 |