missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-methylpyridine, 95%, Thermo Scientific Chemicals

Catalog Number p-7053965
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
39621-00-6
C6H5Cl2N
162.013
MFCD09264302
GTHRJKYVJZJPCF-UHFFFAOYSA-N
10898997
2,6-dichloro-4-methylpyridine
CC1=CC(=NC(=C1)Cl)Cl
This item is not returnable. View return policy
39621-00-6
C6H5Cl2N
162.013
MFCD09264302
GTHRJKYVJZJPCF-UHFFFAOYSA-N
10898997
2,6-dichloro-4-methylpyridine
CC1=CC(=NC(=C1)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Synonym 2,6-dichloro-4-picoline, 2,6-dichloro-4-methyl-pyridine, pyridine, 2,6-dichloro-4-methyl, pubchem5534, acmc-209j6i, pydc11, pyridine,2,6-dichloro-4-methyl, 2,6-bis chloranyl-4-methyl-pyridine
Formula Weight 162.02
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2,6-Dichloro-4-methylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'