Learn More
2,6-Dichloro-4-nitraminopyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Dichloro-4-nitraminopyridine |
| CAS | 2587-03-3 |
| Molecular Formula | C5 H3 Cl2 N3 O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C5H3Cl2N3O2/c6-4-1-3(9-10(11)12)2-5(7)8-4/h1-2H,(H,8,9) |
| SMILES | [O-][N+](=O)Nc1cc(Cl)nc(Cl)c1 |
| IUPAC Name | N-(2,6-dichloropyridin-4-yl)nitramide |
| Molecular Weight (g/mol) | 208 |
| Formula Weight | 206.9602 |