missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-(trifluoromethoxy)aniline, 96%, Thermo Scientific™

Catalog Number 11985221
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
99479-66-0
C7H4Cl2F3NO
246.01
MFCD00190128
FKISQWQHZULEEG-UHFFFAOYSA-N
2,6-dichloro-4-trifluoromethoxy aniline, benzenamine, 2,6-dichloro-4-trifluoromethoxy, 2,6-dichloro-4-trifluoromethoxy phenylamine, pubchem8461, acmc-209scs, 3.5-dichloro-4-amino trifluoromethoxybenzene, 2,6-dichloro-4-trifluoromethoxy-phenylamine, 3,5-dichloro-4-amino trifluoromethoxybenzene, 3,5-dichloro-4-aminotrifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy benzenamine
688247
2,6-dichloro-4-(trifluoromethoxy)aniline
NC1=C(Cl)C=C(OC(F)(F)F)C=C1Cl
This item is not returnable. View return policy
99479-66-0
C7H4Cl2F3NO
246.01
MFCD00190128
FKISQWQHZULEEG-UHFFFAOYSA-N
2,6-dichloro-4-trifluoromethoxy aniline, benzenamine, 2,6-dichloro-4-trifluoromethoxy, 2,6-dichloro-4-trifluoromethoxy phenylamine, pubchem8461, acmc-209scs, 3.5-dichloro-4-amino trifluoromethoxybenzene, 2,6-dichloro-4-trifluoromethoxy-phenylamine, 3,5-dichloro-4-amino trifluoromethoxybenzene, 3,5-dichloro-4-aminotrifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy benzenamine
688247
2,6-dichloro-4-(trifluoromethoxy)aniline
NC1=C(Cl)C=C(OC(F)(F)F)C=C1Cl
CAS Min % 95.0
CAS Max % 100.0
Melting Point 32°C to 34°C
Color Brown to Colorless
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 246.02
Percent Purity 96%
Physical Form Powder, Low Melting Solid or Crystalline Needles
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethoxy)aniline
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dichloro-4-(trifluoromethoxy)aniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'