Learn More
2,6-Dichloro-N-(4-chlorophenyl)-benzeneacetamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Dichloro-N-(4-chlorophenyl)-benzeneacetamide |
| CAS | 560075-65-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 H10 Cl3 N O |
| Quantity | 5 g |
| Synonym | 2,6-Dichloro-N-(4-chlorophenyl)benzeneacetamide, Diclofenac RC F (USP), Imp F (EP) |
| InChI Formula | InChI=1S/C14H10Cl3NO/c15-9-4-6-10(7-5-9)18-14(19)8-11-12(16)2-1-3-13(11)17/h1-7H,8H2,(H,18,19) |
| SMILES | Clc1ccc(NC(=O)Cc2c(Cl)cccc2Cl)cc1 |
| IUPAC Name | N-(4-chlorophenyl)-2-(2,6-dichlorophenyl)acetamide |
| Molecular Weight (g/mol) | 314.59 |
| Show More |