Learn More
2,6-Dichloroterephthalic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Dichloroterephthalic acid |
| CAS | 116802-97-2 |
| Molecular Formula | C8 H4 Cl2 O4 |
| Quantity | 50 mg |
| Synonym | 1,4-Benzenedicarboxylic acid, 2,6-dichloro-, 2,6-Dichloro-1,4-benzenedicarboxylic acid, 2,6-Dichloroterephthalic acid |
| InChI Formula | InChI=1S/C8H4Cl2O4/c9-4-1-3(7(11)12)2-5(10)6(4)8(13)14/h1-2H,(H,11,12)(H,13,14) |
| SMILES | OC(=O)c1cc(Cl)c(C(=O)O)c(Cl)c1 |
| IUPAC Name | 2,6-dichloroterephthalic acid |
| Molecular Weight (g/mol) | 235.02 |
| Formula Weight | 233.9487 |