missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15474866
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474866 1 g 1g
15484866 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474866 Supplier Thermo Scientific Alfa Aesar Supplier No. H59195.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1218791-32-2
Molecular Formula C14H17BF2O4
Molecular Weight (g/mol) 298.093
MDL Number MFCD12546491
InChI Key CGADBVGBEFYTMK-UHFFFAOYSA-N
Synonym methyl 3,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid, pinacol ester, 2,6-difluoro-4-methoxycarbonyl benzeneboronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenyl boronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid,pinacol ester, methyl 3,5-difluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, methyl3,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
PubChem CID 46739549
IUPAC Name methyl 3,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2F)C(=O)OC)F

Spezifikation

Quantity 1 g
Formula Weight 298.09
Percent Purity 96%
Chemical Name or Material 2,6-Difluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.