Learn More
2,6-Dimethylquinoline, 98%
Product Code. 11416806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416806 5 g 5g
11426806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11416806 Supplier Thermo Scientific Alfa Aesar Supplier No. B23934.06
Request Bulk or Custom Format
CAS: 877-43-0 | C11H11N | 157.216 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 877-43-0
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.216
MDL Number MFCD00006762
InChI Key JJPSZKIOGBRMHK-UHFFFAOYSA-N
Synonym p-toluquinaldine, quinoline, 2,6-dimethyl, 6-methylquinaldine, unii-kst0m1t4mb, 2,6-dimethyl-quinoline, kst0m1t4mb, pubchem5876, quinoline,6-dimethyl, acmc-209qpt, 2,6-dimethylquinoline;
PubChem CID 13414
IUPAC Name 2,6-dimethylquinoline
SMILES CC1=CC2=C(C=C1)N=C(C=C2)C
Specifications
Melting Point 56°C to 58°C
Boiling Point 266°C to 267°C
Quantity 5 g
Formula Weight 157.21
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylquinoline