missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylquinoline, 98%

Product Code. 11426806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426806 25 g 25g
11416806 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426806 Supplier Thermo Scientific Alfa Aesar Supplier No. B23934.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 877-43-0 | C11H11N | 157.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-43-0
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.216
MDL Number MFCD00006762
InChI Key JJPSZKIOGBRMHK-UHFFFAOYSA-N
Synonym p-toluquinaldine, quinoline, 2,6-dimethyl, 6-methylquinaldine, unii-kst0m1t4mb, 2,6-dimethyl-quinoline, kst0m1t4mb, pubchem5876, quinoline,6-dimethyl, acmc-209qpt, 2,6-dimethylquinoline;
PubChem CID 13414
IUPAC Name 2,6-dimethylquinoline
SMILES CC1=CC2=C(C=C1)N=C(C=C2)C

Specifications

Melting Point 56°C to 58°C
Boiling Point 266°C to 267°C
Quantity 25 g
Formula Weight 157.21
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylquinoline
compliance-icons
Product Identifier
  • 2,6-Dimethylquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2,6-Dimethylquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.