missing translation for 'onlineSavingsMsg'
Learn More

2-Acrylamido-2-methylpropanesulfonic acid, 98%

Product Code. p-7024376
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11474040

Brand: Thermo Scientific Alfa Aesar A18924.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15214-89-8 | C7H13NO4S | 207.24 g/mol

2-Acrylamide-2-methylpropanesulfonic acid has good complexion, adsorption, biological activity, surface activity, hydrolysis stability and thermal stability. It can be used in oil chemical, water treatment, synthetic fiber, printing and dyeing, plastics, water absorbing coatings, paper, bio-medical, magnetic materials and cosmetics industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acrylamide-2-methylpropanesulfonic acid has good complexion, adsorption, biological activity, surface activity, hydrolysis stability and thermal stability. It can be used in oil chemical, water treatment, synthetic fiber, printing and dyeing, plastics, water absorbing coatings, paper, bio-medical, magnetic materials and cosmetics industries.

Solubility
Highly soluble in water (1×106 mg/L at 25°C ).

Notes
Hygroscopic. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15214-89-8
Molecular Formula C7H13NO4S
Molecular Weight (g/mol) 207.24
MDL Number MFCD00007522
InChI Key XHZPRMZZQOIPDS-UHFFFAOYSA-N
Synonym 2-acrylamido-2-methylpropanesulfonic acid, 2-acrylamido-2-methyl-1-propanesulfonic acid, 2-acrylamide-2-methylpropanesulfonic acid, 1-propanesulfonic acid, 2-methyl-2-1-oxo-2-propenyl amino, polyacrylamidomethylpropane sulfonic acid, unii-490hqe5ki5, 2-acrylamido-2-methylpropanesulfonate, 2-acrylamido-2-methylpropanesulphonic acid, 2-acrylamido-2-methylpropane sulfonic acid, 2-acrylamido-2-methylpropane-1-sulfonic acid
PubChem CID 65360
IUPAC Name 2-methyl-2-(prop-2-enoylamino)propane-1-sulfonic acid
SMILES CC(C)(CS(O)(=O)=O)NC(=O)C=C

Specifications

Melting Point ∼195°C (decomposition)
Quantity 50 g
UN Number UN2585
Beilstein 1946464
Formula Weight 207.25
Percent Purity 98%
Chemical Name or Material 2-Acrylamido-2-methylpropanesulfonic acid
compliance-icons compliance-icons
Product Identifier
  • 2-Acrylamido-2-methylpropanesulphonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-Acrylamido-2-methylpropanesulphonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.