Learn More
Julabo™ 2-Adapters for Presto™ Temperature Control System
Click to view available options
Connection Type:
M24 x 1.5 F to M24 x 1.5 F
M24 x 1.5 F to M30 x 1.5 F
M24 x 1.5 F to NPT 1 in. F
M24 x 1.5 F to NPT 1/2 in. M
M24 x 1.5 F to NPT 1/4 in. F
M24 x 1.5 F to NPT 1/4 in. M
M24 x 1.5 F to NPT 3/4 in. F
M24 x 1.5 F to NPT 3/4 in. M
M24 x 1.5 F to NPT 3/8 in. F
M24 x 1.5 Female to 1 in. Tube
M24 x 1.5 Female to 1/4 in. Tube
M24 x 1.5 Female to 3/8 in. Tube
M24 x 1.5 Female to M30 x 1.5 Male
M24 x 1.5 Female to NPT 1 in. Male
M24 x 1.5 Male to M16 x 1.5 Female
M24 x 1.5 Male to M30 x 1.5 Female
M30 x 1.5 F to M30 x 1.5 F
M30 x 1.5 F to M38 x 1.5 F
M30 x 1.5 F to NPT 1 in. F
M30 x 1.5 F to NPT 3/4 in. F
M30 x 1.5 F to NPT 3/4 in. M
M30 x 1.5 Female to 1 in. Tube
M30 x 1.5 Female to M38 x 1.5 Male
M30 x 1.5 Female to NPT 1 in. Male
M30 x 1.5 Male to M38 x 1.5 Female
M30 x 1.5 Male to Male
M38 x 1.5 F to M38 x 1.5 F
M38 x 1.5 F to NPT 1 in. F
M38 x 1.5 F to NPT1 1/4 in. F
M38 x 1.5 F to NPT1 1/4 in. M
M38 x 1.5 Female to 1 in. Tube
M38 x 1.5 Female to NPT 1 in. Male
M38 x 1.5 Male to Male
NPT 3/4 in. Female to M16 x 1 Male
Unit Size:
Pack of 2
Specifications
Specifications
| For Use With (Equipment) | Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System |
| Type | 2-Adapters |
| Connection Type | M24 x 1.5 Male to M16 x 1.5 Female |