Learn More
- Home
- Products
- Baths
- Bath Accessories
- Julabo™ 2-Adapters for Presto™ Temperature Control System
Julabo™ 2-Adapters for Presto™ Temperature Control System
Product Code. 15294263
Change view
Click to view available options
Connection Type:
M24 x 1.5 F to M24 x 1.5 F
M24 x 1.5 F to M30 x 1.5 F
M24 x 1.5 F to NPT 1 in. F
M24 x 1.5 F to NPT 1/2 in. M
M24 x 1.5 F to NPT 1/4 in. F
M24 x 1.5 F to NPT 1/4 in. M
M24 x 1.5 F to NPT 3/4 in. F
M24 x 1.5 F to NPT 3/4 in. M
M24 x 1.5 F to NPT 3/8 in. F
M24 x 1.5 Female to 1 in. Tube
M24 x 1.5 Female to 1/4 in. Tube
M24 x 1.5 Female to 3/8 in. Tube
M24 x 1.5 Female to M30 x 1.5 Male
M24 x 1.5 Female to NPT 1 in. Male
M24 x 1.5 Male to M16 x 1.5 Female
M24 x 1.5 Male to M30 x 1.5 Female
M30 x 1.5 F to M30 x 1.5 F
M30 x 1.5 F to M38 x 1.5 F
M30 x 1.5 F to NPT 1 in. F
M30 x 1.5 F to NPT 3/4 in. F
M30 x 1.5 F to NPT 3/4 in. M
M30 x 1.5 Female to 1 in. Tube
M30 x 1.5 Female to M38 x 1.5 Male
M30 x 1.5 Female to NPT 1 in. Male
M30 x 1.5 Male to M38 x 1.5 Female
M30 x 1.5 Male to Male
M38 x 1.5 F to M38 x 1.5 F
M38 x 1.5 F to NPT 1 in. F
M38 x 1.5 F to NPT1 1/4 in. F
M38 x 1.5 F to NPT1 1/4 in. M
M38 x 1.5 Female to 1 in. Tube
M38 x 1.5 Female to NPT 1 in. Male
M38 x 1.5 Male to Male
NPT 3/4 in. Female to M16 x 1 Male
Unit Size:
Pack of 2
34 product options available for selection
| Product Code. | Connection Type | unitSize |
|---|---|---|
| 15294263 | M24 x 1.5 Female to NPT 1 in. Male | Pack of 2 |
| 15224273 | M24 x 1.5 Female to 1 in. Tube | Pack of 2 |
| 15204273 | M24 x 1.5 Female to 1/4 in. Tube | Pack of 2 |
| 15214273 | M24 x 1.5 Female to 3/8 in. Tube | Pack of 2 |
| 15234273 | M24 x 1.5 F to M24 x 1.5 F | Pack of 2 |
| 15234263 | M24 x 1.5 F to NPT 1 in. F | Pack of 2 |
| 15264263 | M24 x 1.5 F to NPT 1/2 in. M | Pack of 2 |
| 15244263 | M24 x 1.5 F to NPT 1/4 in. M | Pack of 2 |
| 15244273 | M24 x 1.5 Female to M30 x 1.5 Male | Pack of 2 |
| 15294253 | M24 x 1.5 F to NPT 1/4 in. F | Pack of 2 |
| 15264273 | M24 x 1.5 F to M30 x 1.5 F | Pack of 2 |
| 15214263 | M24 x 1.5 F to NPT 3/4 in. F | Pack of 2 |
| 15274263 | M24 x 1.5 F to NPT 3/4 in. M | Pack of 2 |
| 15204263 | M24 x 1.5 F to NPT 3/8 in. F | Pack of 2 |
| 15274273 | M24 x 1.5 Male to M16 x 1.5 Female | Pack of 2 |
| 15254273 | M24 x 1.5 Male to M30 x 1.5 Female | Pack of 2 |
| 15264283 | M30 x 1.5 Female to 1 in. Tube | Pack of 2 |
| 15204283 | M30 x 1.5 F to M38 x 1.5 F | Pack of 2 |
| 15274283 | M30 x 1.5 F to M30 x 1.5 F | Pack of 2 |
| 15284273 | M30 x 1.5 Female to M38 x 1.5 Male | Pack of 2 |
| 15254283 | M30 x 1.5 F to NPT 1 in. F | Pack of 2 |
| 15234283 | M30 x 1.5 Female to NPT 1 in. Male | Pack of 2 |
| 15224283 | M30 x 1.5 F to NPT 3/4 in. F | Pack of 2 |
| 15294273 | M30 x 1.5 Male to M38 x 1.5 Female | Pack of 2 |
| 15214283 | M30 x 1.5 F to NPT 3/4 in. M | Pack of 2 |
| 15254293 | M30 x 1.5 Male to Male | Pack of 2 |
| 15234293 | M38 x 1.5 Female to 1 in. Tube | Pack of 2 |
| 15284283 | M38 x 1.5 F to M38 x 1.5 F | Pack of 2 |
| 15204293 | M38 x 1.5 F to NPT 1 in. F | Pack of 2 |
| 15294283 | M38 x 1.5 Female to NPT 1 in. Male | Pack of 2 |
| 15224293 | M38 x 1.5 F to NPT1 1/4 in. F | Pack of 2 |
| 15214293 | M38 x 1.5 F to NPT1 1/4 in. M | Pack of 2 |
| 15264293 | M38 x 1.5 Male to Male | Pack of 2 |
| 11954718 | NPT 3/4 in. Female to M16 x 1 Male | Pack of 2 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 34 options available.
Specifications
| For Use With (Equipment) | Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System |
| Type | 2-Adapters |
| Connection Type | M24 x 1.5 Female to NPT 1 in. Male |