Learn More
2-Allyl-N-Boc-L-glycine dicyclohexylamine salt, 95%, Thermo Scientific™
Product Code. p-7034379
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15479545
Brand: Thermo Scientific Alfa Aesar H52062.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Allyl-N-Boc-L-glycine dicyclohexylamine salt is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 143979-15-1
Molecular Formula C22H40N2O4
Molecular Weight (g/mol) 396.572
MDL Number MFCD01321013
InChI Key VMCGMPITVQIMGK-ZLTKDMPESA-N
Synonym dicyclohexylamine s-2-tert-butoxycarbonyl amino pent-4-enoate, boc-l-2-allylglycine dicyclohexylamine salt, boc-l-allylglycine.dcha, boc-l-allylglycine-dcha, boc-l-allylglycine dicyclohexylaminoium salt, boc-d-allylglycine-dcha, 2s-2-tert-butoxycarbonyl amino pent-4-enoic acid; dicha, boc-l-allylglycine dcha, boc-algly-oh dcha, boc-l-allyglycine?cha
PubChem CID 2755982
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
SMILES CC(C)(C)OC(=O)NC(CC=C)C(=O)O.C1CCC(CC1)NC2CCCCC2
Specifications
Quantity 250 mg
Formula Weight 396.57
Percent Purity 95%
Chemical Name or Material 2-Allyl-N-Boc-L-glycine dicyclohexylamine salt