missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-(4-morpholinyl)-ethanone Hydrochloride, TRC

Product Code. 30325820
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325820 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325820 Supplier Toronto Research Chemicals Supplier No. TRCA618713250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-1-(4-morpholinyl)-ethanone Hydrochloride
CAS 24152-96-3
Molecular Formula C6H12N2O2.HCl
Quantity 250 mg
Synonym 2-Amino-1-(4-morpholinyl)-ethanone Hydrochloride
InChI Formula InChI=1S/C6H12N2O2.ClH/c7-5-6(9)8-1-3-10-4-2-8;/h1-5,7H2;1H
SMILES NCC(N1CCOCC1)=O.[H]Cl
IUPAC Name 2-Amino-1-(4-morpholinyl)-ethanone Hydrochloride
Molecular Weight (g/mol) 180.63
Formula Weight 180.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.