missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-benzyloxypyrazine, 96%, Thermo Scientific Chemicals

Catalog Number p-7054758
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
110223-15-9
C11H11N3O
201.23
MFCD09838954
QKEQFJXWHGVJJU-UHFFFAOYSA-N
2-amino-3-benzyloxypyrazine, 3-benzyloxy pyrazin-2-amine, 2-pyrazinamine,3-phenylmethoxy, 2-pyrazinamine, 3-phenylmethoxy, zlchem 177, pubchem19570, acmc-20a0ri, 2-aminol-3-benzyloxypyrazine, 3-phenylmethoxy-2-pyrazinamine
13900234
3-phenylmethoxypyrazin-2-amine
NC1=NC=CN=C1OCC1=CC=CC=C1
This item is not returnable. View return policy
110223-15-9
C11H11N3O
201.23
MFCD09838954
QKEQFJXWHGVJJU-UHFFFAOYSA-N
2-amino-3-benzyloxypyrazine, 3-benzyloxy pyrazin-2-amine, 2-pyrazinamine,3-phenylmethoxy, 2-pyrazinamine, 3-phenylmethoxy, zlchem 177, pubchem19570, acmc-20a0ri, 2-aminol-3-benzyloxypyrazine, 3-phenylmethoxy-2-pyrazinamine
13900234
3-phenylmethoxypyrazin-2-amine
NC1=NC=CN=C1OCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 201.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-3-benzyloxypyrazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'