missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-benzyloxypyridine-5-boronic acid pinacol ester, 96%

Product Code. 15441066
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15441066

Brand: Thermo Scientific Alfa Aesar H54341.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 756520-57-7 | C18H23BN2O3 | 326.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 756520-57-7
Molecular Formula C18H23BN2O3
Molecular Weight (g/mol) 326.203
MDL Number MFCD13191381
InChI Key RPQADWBEACHUFT-UHFFFAOYSA-N
Synonym 2-amino-3-benzyloxypyridine-5-boronic acid pinacol ester, 3-benzyloxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine
PubChem CID 21111425
IUPAC Name 3-phenylmethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)OCC3=CC=CC=C3

Specifications

Melting Point 135°C to 137°C
Quantity 250 mg
Formula Weight 326.2
Percent Purity 96%
Chemical Name or Material 2-Amino-3-benzyloxypyridine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.