Learn More
2-Amino-3-fluoropyridine, 97%
Product Code. 15474617
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474617 5 g 5g
15464617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15474617 Supplier Thermo Scientific Alfa Aesar Supplier No. H31970.06
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
CAS: 21717-95-3 | C5H5FN2 | 112.11 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21717-95-3
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.11
MDL Number MFCD04114135
InChI Key WWEINXQNCAWBPD-UHFFFAOYSA-N
Synonym 2-amino-3-fluoropyridine, 3-fluoro-pyridin-2-ylamine, 2-pyridinamine, 3-fluoro, 3-fluoro-2-pyridylamine, 3-fluoro-2-pyridinamine, 3-fluoro-2-aminopyridine, fluoropyridimine, pubchem2976, acmc-209fng, 3-fluoro-pyridin-2ylamine
PubChem CID 2762809
IUPAC Name 3-fluoropyridin-2-amine
SMILES NC1=C(F)C=CC=N1
Specifications
Melting Point 39°C to 43°C
Odor Characteristic
Quantity 5 g
UN Number UN2811
Formula Weight 112.11
Percent Purity 97%
Chemical Name or Material 2-Amino-3-fluoropyridine