Learn More
2-Amino-4-methoxypyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Amino-4-methoxypyridine |
| CAS | 10201-73-7 |
| Molecular Formula | C6H8N2O |
| Quantity | 5 g |
| Synonym | (4-Methoxypyridin-2-yl)amine, 4-(Methyloxy)-2-pyridinamine, 4-Methoxy-2-aminopyridine, 4-Methoxypyridin-2-amine |
| InChI Formula | InChI=1S/C6H8N2O/c1-9-5-2-3-8-6(7)4-5/h2-4H,1H3,(H2,7,8) |
| SMILES | COc1ccnc(N)c1 |
| IUPAC Name | 4-methoxypyridin-2-amine |
| Molecular Weight (g/mol) | 124.14 |
| Formula Weight | 124.0637 |