Learn More
2-amino-4-picoline, 98%, Thermo Scientific Chemicals
Catalog Number p-3735517
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
695-34-1
C6H8N2
108.14
MFCD00006321
ORLGLBZRQYOWNA-UHFFFAOYSA-N
2-amino-4-methylpyridine, 2-amino-4-picoline, 2-pyridinamine, 4-methyl, ascensil, 4-methyl-2-pyridinamine, 4-methyl-2-pyridylamine, 4-methyl-2-aminopyridine, 4-picoline, 2-amino, w 45 raschig, aminton
1533
4-methylpyridin-2-amine
CC1=CC(=NC=C1)N
695-34-1
C6H8N2
108.14
MFCD00006321
ORLGLBZRQYOWNA-UHFFFAOYSA-N
2-amino-4-methylpyridine, 2-amino-4-picoline, 2-pyridinamine, 4-methyl, ascensil, 4-methyl-2-pyridinamine, 4-methyl-2-pyridylamine, 4-methyl-2-aminopyridine, 4-picoline, 2-amino, w 45 raschig, aminton
1533
4-methylpyridin-2-amine
CC1=CC(=NC=C1)N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 96°C to 101°C
Color Beige to Yellow or Brown
Boiling Point 230°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 22, V,9, 325
Merck Index 15, 462
Solubility Information Solubility in water: 41g/L (20°C). Other solubilities: soluble in lower alcohols,slightly soluble in petroleum ether
Formula Weight 108.14
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material 2-Amino-4-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-4-picoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.