Learn More
2-Amino-5-fluorobenzotrifluoride, 98%, BLD Pharm™
Product Code. 30987644
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30987644 5 g
30973422 25 g
30978348 100 g
30978783 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30987644 Supplier BLD Pharm Supplier No. BD103655g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 393-39-5
Molecular Formula C7H5F4N
MDL Number MFCD00007831
Quantity 5 g
SMILES NC1=CC=C(F)C=C1C(F)(F)F
Molecular Weight (g/mol) 179.11
Percent Purity 98%
Chemical Name or Material 2-Amino-5-fluorobenzotrifluoride