missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloro-3-nitropyridine, 97%, Thermo Scientific™

Catalog Number p-7077187
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
27048-04-0
C5H4ClN3O2
173.56
MFCD00044350
WERABQRUGJIMKQ-UHFFFAOYSA-N
2-amino-6-chloro-3-nitropyridine, 2-amino-3-nitro-6-chloropyridine, 6-chloro-3-nitro-2-pyridylamine, 6-chloro-3-nitro-2-pyridinamine, 6-amino-2-chloro-5-nitropyridine, 6-chloro-3-nitropyridin-2-ylamine, 6-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 6-chloro-3-nitro, 6-chloro-3-nitro-pyridin-2-amine, zlchem 113
2724035
6-chloro-3-nitropyridin-2-amine
NC1=NC(Cl)=CC=C1[N+]([O-])=O
This item is not returnable. View return policy
27048-04-0
C5H4ClN3O2
173.56
MFCD00044350
WERABQRUGJIMKQ-UHFFFAOYSA-N
2-amino-6-chloro-3-nitropyridine, 2-amino-3-nitro-6-chloropyridine, 6-chloro-3-nitro-2-pyridylamine, 6-chloro-3-nitro-2-pyridinamine, 6-amino-2-chloro-5-nitropyridine, 6-chloro-3-nitropyridin-2-ylamine, 6-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 6-chloro-3-nitro, 6-chloro-3-nitro-pyridin-2-amine, zlchem 113
2724035
6-chloro-3-nitropyridin-2-amine
NC1=NC(Cl)=CC=C1[N+]([O-])=O
Melting Point 193.0°C to 198.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 173.56
Percent Purity 97%
Chemical Name or Material 2-Amino-6-chloro-3-nitropyridine
compliance-icons
Product Identifier
  • 2-Amino-6-chloro-3-nitropyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'