Learn More
2-Amino-6-(chloromethyl)-4(3H)-pteridinone (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 6-(Chloromethyl)pterin |
| CAS | 873397-19-4 |
| Molecular Formula | C7 H6 Cl N5 O |
| Quantity | 10 mg |
| Synonym | 4(3H)-Pteridinone, 2-amino-6-(chloromethyl)-, 4-Pteridinol, 2-amino-6-(chloromethyl)- (5CI), 2-Amino-6-(chloromethyl)-4(3H)-pteridinone, 6-(Chloromethyl)pterin |
| InChI Formula | InChI=1S/C7H6ClN5O/c8-1-3-2-10-5-4(11-3)6(14)13-7(9)12-5/h2H,1H2,(H3,9,10,12,13,14) |
| SMILES | NC1=NC(=O)c2nc(CCl)cnc2N1 |
| IUPAC Name | 2-amino-6-(chloromethyl)-1H-pteridin-4-one |
| Molecular Weight (g/mol) | 211.61 |
| Formula Weight | 211.0261 |