missing translation for 'onlineSavingsMsg'
Learn More

2-amino-6-methylbenzamide, TRC

Product Code. 30315135
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315135 50 mg 50mg
30312435 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30315135 Supplier Toronto Research Chemicals Supplier No. TRCA61847850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-amino-6-methylbenzamide
CAS 1885-31-0
Molecular Formula C8H10N2O
Quantity 50 mg
InChI Formula InChI=1S/C8H10N2O/c1-5-3-2-4-6(9)7(5)8(10)11/h2-4H,9H2,1H3,(H2,10,11)
SMILES Cc1cccc(N)c1C(=O)N
IUPAC Name 2-amino-6-methylbenzamide
Molecular Weight (g/mol) 150.17
Formula Weight 150.0793
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.