missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-a-(methoxyimino)-4-thiazoleacetic Acid, TRC

Product Code. 30328478
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328478 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328478 Supplier Toronto Research Chemicals Supplier No. TRCA61196525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2Z)-(2-Aminothiazol-4-yl)(methoxyimino)acetic Acid
CAS 65872-41-5
Molecular Formula C6 H7 N3 O3 S
Quantity 25 g
Synonym 4-Thiazoleacetic acid, 2-amino-α-(methoxyimino)-, (αZ)-, 4-Thiazoleacetic acid, 2-amino-α-(methoxyimino)-, (Z)-, (Z)-(2-Aminothiazol-4-yl)-α-(methoxyimino)acetic acid, (Z)-(2-Aminothiazol-4-yl)methoxyiminoacetic acid, (Z)-2-(2-Aminothiazol-4-yl)-2-methoxyiminoacetic acid, (Z)-2-Amino-α-(methoxyimino)-4-thiazoleacetic acid, (Z)-2-Amino-α-(methoxyimino)-4-thiazoleacetic acid, 2-(2-Amino-4-thiazolyl)-(Z)-2-methoxyiminoacetic acid, 2-(2-Aminothiazol-4-yl)-2-(Z)-methoxyiminoacetic acid, 2-(2-Aminothiazole-4-yl)-2-(Z)-methoxyiminoacetic acid, Thiazoximic acid, syn-2-(2-Aminothiazol-4-yl)-2-methoxyiminoacetic acid, syn-Thiazoximic acid, (2Z)-(2-Aminothiazol-4-yl)(methoxyimino)acetic acid
InChI Formula InChI=1S/C6H7N3O3S/c1-12-9-4(5(10)11)3-2-13-6(7)8-3/h2H,1H3,(H2,7,8)(H,10,11)/b9-4-
SMILES CO\N=C(/C(=O)O)\c1csc(N)n1
IUPAC Name (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetic acid
Molecular Weight (g/mol) 201.2
Formula Weight 201.0208
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.