Learn More
2-Aminoethanethiol hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 156-57-0 |
| Molecular Formula | C2H8ClNS |
| MDL Number | MFCD00012904 |
| Quantity | 500 g |
| SMILES | NCCS.[H]Cl |
| Molecular Weight (g/mol) | 113.61 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Aminoethanethiol hydrochloride |