missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoindan hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-4229061
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2338-18-3
C9H11ClN
168.64
MFCD00012549
DLYRFJDJKGGVKT-UHFFFAOYSA-N
2-aminoindan hydrochloride, 2-aminoindane hydrochloride, 2,3-dihydro-1h-inden-2-amine hydrochloride, 2-aminoindane hcl, 2-indanamine hydrochloride, 2-indanamine, hydrochloride, 2-indanylamine hydrochloride, su 8629 hydrochloride, indan-2-ylammonium chloride, indan-2-ylamine hydrochloride
122764
2,3-dihydro-1H-inden-2-amine;hydrochloride
[Cl].NC1CC2=CC=CC=C2C1
This item is not returnable. View return policy
2338-18-3
C9H11ClN
168.64
MFCD00012549
DLYRFJDJKGGVKT-UHFFFAOYSA-N
2-aminoindan hydrochloride, 2-aminoindane hydrochloride, 2,3-dihydro-1h-inden-2-amine hydrochloride, 2-aminoindane hcl, 2-indanamine hydrochloride, 2-indanamine, hydrochloride, 2-indanylamine hydrochloride, su 8629 hydrochloride, indan-2-ylammonium chloride, indan-2-ylamine hydrochloride
122764
2,3-dihydro-1H-inden-2-amine;hydrochloride
[Cl].NC1CC2=CC=CC=C2C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 246.0°C to 247.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 12,1196
Formula Weight 169.65
Percent Purity 98%
Chemical Name or Material 2-Aminoindan hydrochloride
compliance-icons
Product Identifier
  • 2-Aminoindan hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'