missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[2-(Benzyloxy)-2-oxoethyl](4-phenylbutyl)phosphinic Acid, Mikromol™
Description
[2-(Benzyloxy)-2-oxoethyl](4-phenylbutyl)phosphinic Acid
Specifications
Specifications
| Chemical Name or Material | [2-(Benzyloxy)-2-oxoethyl](4-phenylbutyl)phosphinic Acid |
| CAS | 87460-09-1 |
| Analyte or Component Names | [2-(Benzyloxy)-2-oxoethyl](4-phenylbutyl)phosphinic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H23 O4 P |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-[Hydroxy(4-phenylbutyl)phosphinyl]acetic Acid 1-(Phenylmethyl) Ester |
| InChI Formula | InChI=1S/C19H23O4P/c20-19(23-15-18-12-5-2-6-13-18)16-24(21,22)14-8-7-11-17-9-3-1-4-10-17/h1-6,9-10,12-13H,7-8,11,14-16H2,(H,21,22) |
| SMILES | OP(=O)(CCCCc1ccccc1)CC(=O)OCc2ccccc2 |
| IUPAC Name | (2-benzyloxy-2-oxo-ethyl)-(4-phenylbutyl)phosphinic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?