Learn More
2-Benzylthio-5-amino-1,3,4-thiadiazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Benzylthio-5-amino-1,3,4-thiadiazole |
| CAS | 25660-71-3 |
| Molecular Formula | C8 H5 N3 O3 |
| Quantity | 250 mg |
| Synonym | 5-[(Phenylmethyl)thio]-1,3,4-thiadiazol-2-amine, 2-Amino-5-benzylmercapto-1,3,4-thiadiazole, 2-Amino-5-benzylthio-1,2,4-thiadiazol, 2-Amino-5-benzylthio-1,3,4-thiadiazole, 5-Amino-2-(benzylthio)-1,3,4-thiadiazole, 5-Benzylsulfanyl-2-amino-1,3,4-thiadiazole, NSC 204491 |
| InChI Formula | InChI=1S/C8H5N3O3/c12-8-6-3-5(11(13)14)1-2-7(6)9-4-10-8/h1-4H,(H,9,10,12) |
| SMILES | [O-][N+](=O)c1ccc2N=CNC(=O)c2c1 |
| IUPAC Name | 6-nitro-3H-quinazolin-4-one |
| Molecular Weight (g/mol) | 191.144 |
| Formula Weight | 191.033 |