missing translation for 'onlineSavingsMsg'
Learn More

2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde, 97%, Thermo Scientific™

Catalog Number 15448136
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
669091-00-3
C23H23OP
346.41
MFCD15143662
PQIAXRGPKDEYLW-UHFFFAOYSA-N
2-bis 3,5-dimethylphenyl phosphino benzaldehyde, 2-bis 3,5-dimethylphenyl phosphanyl benzaldehyde, acmc-20ao6m, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde,min
71310677
2-bis(3,5-dimethylphenyl)phosphanylbenzaldehyde
CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=CC=C1C=O
This item is not returnable. View return policy
669091-00-3
C23H23OP
346.41
MFCD15143662
PQIAXRGPKDEYLW-UHFFFAOYSA-N
2-bis 3,5-dimethylphenyl phosphino benzaldehyde, 2-bis 3,5-dimethylphenyl phosphanyl benzaldehyde, acmc-20ao6m, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde,min
71310677
2-bis(3,5-dimethylphenyl)phosphanylbenzaldehyde
CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=CC=C1C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 106°C to 110°C
Color Yellow
Quantity 500 mg
Sensitivity Air Sensitive
Formula Weight 346.4
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'