missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)ethyl bromide, 96%

Product Code. 15491378
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15491378

Brand: Thermo Scientific Alfa Aesar H55211.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 39684-80-5 | C7H14BrNO2 | 224.10 g/mol

N-t-Boc-2-bromoethylamine is a building block for preparing fluorinated spacers having nucleophilic and electrophilic termini.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-t-Boc-2-bromoethylamine is a building block for preparing fluorinated spacers having nucleophilic and electrophilic termini.

Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store at -20°C. Reactive with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39684-80-5
Molecular Formula C7H14BrNO2
Molecular Weight (g/mol) 224.10
MDL Number MFCD02683428
InChI Key TZRQZPMQUXEZMC-UHFFFAOYSA-N
Synonym tert-butyl n-2-bromoethyl carbamate, 2-boc-amino ethyl bromide, n-boc-bromoethylamine, tert-butyl 2-bromoethyl carbamate, n-boc-2-bromoethylamine, tert-butyl 2-bromoethylcarbamate, 2-bromo-ethyl-carbamic acid tert-butyl ester, 2-tert-butoxycarbonylamino ethyl bromide, 2-boc-amino-ethyl bromide
PubChem CID 4103526
IUPAC Name tert-butyl N-(2-bromoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCBr

Specifications

Melting Point 30°C to 32°C
Linear Formula BrCH2CH2NHCO2C(CH3)3
Beilstein 2325117
Solubility Information Soluble in Chloroform,Dichloromethane and Ethyl Acetate.
Quantity 250 mg
Formula Weight 224.1
Percent Purity 96%
Chemical Name or Material 2-(Boc-amino)ethyl bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.