missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 97%

Product Code. 11905331 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11905331 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11905331 Supplier Thermo Scientific Acros Supplier No. 439960050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 878-23-9 | C7H9BBrF4N | 273.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 878-23-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 100.0°C to 106.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Molecular Formula C7H9BBrF4N
MDL Number MFCD00060046
Quantity 5 g
Synonym 2-bromo-1-ethylpyridinium tetrafluoroborate, 2-bromo-1-ethylpyridin-1-ium tetrafluoroborate, bep tetrafluoroborate, bromoethylpyridiniumtetrafluoroborate, 2-bromo-1-ethyl-pyridinium tetrafluoroborate, bepybr, acmc-209qqc, 2-brom-1-ethyl-pyridiniumtetrafluoroborat, 2-bromo-1-ethyl pyridinium tetrafluoroborate, 2-bromo-1-ethyl-pyridin-1-ium tetrafluoroborate
InChI Key YJDXVQLBIAJTHP-UHFFFAOYSA-N
SMILES F[B-](F)(F)F.CC[N+]1=CC=CC=C1Br
IUPAC Name 2-bromo-1-ethylpyridin-1-ium;tetrafluoroborate
Molecular Weight (g/mol) 273.86
PubChem CID 2736300
Formula Weight 273.86
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1-ethyl-pyridinium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-1-ethylpyridinium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.