Learn More
2-Bromo-2-(2-fluorophenyl)-1-cyclopropylethanone (90% purity), TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-2-(2-fluorophenyl)-1-cyclopropylethanone |
| CAS | 204205-33-4 |
| Molecular Formula | C11H10BrFO |
| Quantity | 1 g |
| Synonym | 2-Bromo-1-cyclopropyl-2-(2-fluorophenyl)ethanone |
| InChI Formula | InChI=1S/C11H10BrFO/c12-10(11(14)7-5-6-7)8-3-1-2-4-9(8)13/h1-4,7,10H,5-6H2 |
| SMILES | FC1=C(C(Br)C(C2CC2)=O)C=CC=C1 |
| IUPAC Name | 2-bromo-1-cyclopropyl-2-(2-fluorophenyl)ethan-1-one |
| Molecular Weight (g/mol) | 257.1 |
| Formula Weight | 255.99 |