missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-difluoroacetophenone, 95%, Thermo Scientific Chemicals

Catalog Number 11974881
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
102429-07-2
C8H5BrF2O
235.03
CSGDTHXBRAAOHV-UHFFFAOYSA-N
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw
2774185
2-bromo-1-(2,4-difluorophenyl)ethanone
C1=CC(=C(C=C1F)F)C(=O)CBr
This item is not returnable. View return policy
102429-07-2
C8H5BrF2O
235.03
CSGDTHXBRAAOHV-UHFFFAOYSA-N
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw
2774185
2-bromo-1-(2,4-difluorophenyl)ethanone
C1=CC(=C(C=C1F)F)C(=O)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 31°C to 33°C
Color White
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 235.03
Percent Purity 95%
Physical Form Low Melting Solid
Chemical Name or Material 2-Bromo-2', 4'-difluoroacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'