Learn More
2-Bromo-3',4'-dichloroacetophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-3',4'-dichloroacetophenone |
| CAS | 2632-10-2 |
| Molecular Formula | C8H5BrCl2O |
| Quantity | 1 g |
| Synonym | 2-Bromo-3',4'-dichloro-acetophenone, 1-(3,4-Dichlorophenyl)-2-bromoethan-1-one, 2-Bromo-1-(3,4-Dichlorophenyl)ethanone, 2-Bromo-3',4'-dichloroacetophenone, 3,4-Dichlorophenacyl Bromide, NSC 55872, alpha-Bromo-3,4-dichloroacetophenone |
| InChI Formula | InChI=1S/C8H5BrCl2O/c9-4-8(12)5-1-2-6(10)7(11)3-5/h1-3H,4H2 |
| SMILES | Clc1ccc(cc1Cl)C(=O)CBr |
| IUPAC Name | 2-bromo-1-(3,4-dichlorophenyl)ethanone |
| Molecular Weight (g/mol) | 267.93 |
| Formula Weight | 265.8901 |